COM638462188 Combi-Blocks, Inc.

N-tert-Butyl-2-chloro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD30527675
  • Cas Number : 2377610-51-8
  • Molecular Weight : 337.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2766
Availability details for
Quantity
  • Availability
  • Add to Cart