COM638462239 Combi-Blocks, Inc.

Memantine

  • MDL number MFCD02114015
  • Cas Number : 19982-08-2
  • Molecular Weight : 179.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-1340
Availability details for
Quantity
  • Availability
  • Add to Cart