COM638462421 Combi-Blocks, Inc.

4-Methyl-2-pentenoic acid

  • MDL number MFCD00043804
  • Cas Number : 10321-71-8
  • Molecular Weight : 114.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2589

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2589
Availability details for
Quantity
  • Availability
  • Add to Cart