COM638462430 Combi-Blocks, Inc.

N-(3-Aminopropyl)cyclohexylamine

  • MDL number MFCD00003838
  • Cas Number : 3312-60-5
  • Molecular Weight : 156.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2647

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2647
Availability details for
Quantity
  • Availability
  • Add to Cart