COM638462743 Combi-Blocks, Inc.

H-Asp(OBzl)-OBzl HCl

  • MDL number MFCD01075101
  • Cas Number : 6327-59-9
  • Molecular Weight : 349.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-8905

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-8905
Availability details for
Quantity
  • Availability
  • Add to Cart