COM638462822 Combi-Blocks, Inc.

3-(2-Methyl-1h-imidazol-1-yl)propanoic acid

  • MDL number MFCD00234356
  • Cas Number : 24647-62-9
  • Molecular Weight : 154.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3187

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3187
Availability details for
Quantity
  • Availability
  • Add to Cart