COM638463132 Combi-Blocks, Inc.

Sodium (3s,4r,5r)-6-((hydrogenphosphonato)oxy)-3,4,5-trihydroxy-2-oxohexyl phosphate

  • MDL number MFCD00135870
  • Cas Number : 81028-91-3
  • Molecular Weight : 428.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5472
Availability details for
Quantity
  • Availability
  • Add to Cart