COM638463358 Combi-Blocks, Inc.

2-(Benzyloxy)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD24039734
  • Cas Number : 1448894-65-2
  • Molecular Weight : 335.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0817

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0817
Availability details for
Quantity
  • Availability
  • Add to Cart