COM638463799 Combi-Blocks, Inc.

2-(2-Aminothiazole-4-yl)-2-methoxyiminoacetic acid

  • MDL number MFCD00071528
  • Cas Number : 65872-41-5
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1329

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1329
Availability details for
Quantity
  • Availability
  • Add to Cart