COM638464050 Combi-Blocks, Inc.

Methyl 1-(triphenylmethyl)-2-aziridinecarboxylate

  • MDL number MFCD09991705
  • Cas Number : 76357-18-1
  • Molecular Weight : 343.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7169

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7169
Availability details for
Quantity
  • Availability
  • Add to Cart