COM638464073 Combi-Blocks, Inc.

Flurbiprofen axetil

  • MDL number MFCD00900049
  • Cas Number : 91503-79-6
  • Molecular Weight : 330.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7645

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7645
Availability details for
Quantity
  • Availability
  • Add to Cart