COM638464126 Combi-Blocks, Inc.

Ethyl 3-oxoquinuclidine-2-carboxylate

  • MDL number MFCD00052371
  • Cas Number : 34286-16-3
  • Molecular Weight : 197.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-oxoquinuclidine-2-carboxylate
Inchi Code 1S/C10H15NO3/c1-2-14-10(13)8-9(12)7-3-5-11(8)6-4-7/h7-8H,2-6H2,1H3
Inchi Key ZJSQPPLVZFFPOW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9080
Availability details for
Quantity
  • Availability
  • Add to Cart