COM638464134 Combi-Blocks, Inc.

3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarboxylic acid

  • MDL number MFCD00137937
  • Cas Number : 55701-05-8
  • Molecular Weight : 209.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9269
Availability details for
Quantity
  • Availability
  • Add to Cart