COM638464249 Combi-Blocks, Inc.

2-Bromothieno[2,3-d]pyridazin-4(5h)-one

  • MDL number MFCD28404699
  • Cas Number : 1936374-21-8
  • Molecular Weight : 231.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3137

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3137
Availability details for
Quantity
  • Availability
  • Add to Cart