COM638464312 Combi-Blocks, Inc.

Ethyl 7-bromo-3-(3-ethoxy-3-oxopropyl)-1h-indole-2-carboxylate

  • MDL number MFCD28405189
  • Cas Number : 1073493-75-0
  • Molecular Weight : 368.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4970

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4970
Availability details for
Quantity
  • Availability
  • Add to Cart