COM638464368 Combi-Blocks, Inc.

3-Amino-2-fluorobenzonitrile

  • MDL number MFCD08062434
  • Cas Number : 873697-68-8
  • Molecular Weight : 136.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-8135

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-8135
Availability details for
Quantity
  • Availability
  • Add to Cart