COM638464434 Combi-Blocks, Inc.

2-(4-Bromophenyl)-2-methylpropane-1,3-diol

  • MDL number MFCD21927427
  • Cas Number : 66810-01-3
  • Molecular Weight : 245.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1541

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1541
Availability details for
Quantity
  • Availability
  • Add to Cart