COM638464483 Combi-Blocks, Inc.

4-Bromo-2-cyanobenzoic acid

  • MDL number MFCD18390738
  • Cas Number : 1223434-16-9
  • Molecular Weight : 226.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4112

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4112
Availability details for
Quantity
  • Availability
  • Add to Cart