COM638464868 Combi-Blocks, Inc.

1,3-Cyclobutanedicarboxylic acid, 1-methyl ester, trans-

  • MDL number MFCD22394024
  • Cas Number : 1401103-71-6
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-8018

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-8018
Availability details for
Quantity
  • Availability
  • Add to Cart