COM638465348 Combi-Blocks, Inc.

(2-Methoxyethyl)benzene

  • MDL number MFCD00025912
  • Cas Number : 3558-60-9
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-0321

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-0321
Availability details for
Quantity
  • Availability
  • Add to Cart