COM638465586 Combi-Blocks, Inc.

Methyl 2-formylthiophene-3-carboxylate

  • MDL number MFCD01859943
  • Cas Number : 24647-82-3
  • Molecular Weight : 170.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-1368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-1368
Availability details for
Quantity
  • Availability
  • Add to Cart