COM638465657 Combi-Blocks, Inc.

Sulfamethazine sodium salt

  • MDL number MFCD00068333
  • Cas Number : 1981-58-4
  • Molecular Weight : 300.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-1672

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-1672
Availability details for
Quantity
  • Availability
  • Add to Cart