COM638465717 Combi-Blocks, Inc.

Trimethylthiourea

  • MDL number MFCD00059148
  • Cas Number : 2489-77-2
  • Molecular Weight : 118.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-1918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-1918
Availability details for
Quantity
  • Availability
  • Add to Cart