COM638465892 Combi-Blocks, Inc.

Raffinose

  • MDL number MFCD00006630
  • Cas Number : 512-69-6
  • Molecular Weight : 504.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-2679

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-2679
Availability details for
Quantity
  • Availability
  • Add to Cart