COM638465975 Combi-Blocks, Inc.

5-Chloro-3-methyl-2(1h)-pyrazinone

  • MDL number MFCD15144356
  • Cas Number : 105985-18-0
  • Molecular Weight : 144.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3011
Availability details for
Quantity
  • Availability
  • Add to Cart