COM638466130 Combi-Blocks, Inc.

3,6-Dimethyl-4-octyn-3,6-diol

  • MDL number MFCD00004480
  • Cas Number : 78-66-0
  • Molecular Weight : 170.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3594

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3594
Availability details for
Quantity
  • Availability
  • Add to Cart