COM638466178 Combi-Blocks, Inc.

Alpha-(2-ethoxyethynyl)-cyclopentanemethanol

  • MDL number MFCD22421705
  • Cas Number : 1038404-02-2
  • Molecular Weight : 168.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3855

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3855
Availability details for
Quantity
  • Availability
  • Add to Cart