COM638466202 Combi-Blocks, Inc.

Methyl 2-propyl-1h-imidazole-5-carboxylate

  • MDL number MFCD29059259
  • Cas Number : 1228376-85-9
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3930

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3930
Availability details for
Quantity
  • Availability
  • Add to Cart