COM638466324 Combi-Blocks, Inc.

Naphthol as

  • MDL number MFCD00004096
  • Cas Number : 92-77-3
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-4464

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-4464
Availability details for
Quantity
  • Availability
  • Add to Cart