COM638466548 Combi-Blocks, Inc.

Bis(2-ethylhexyl) 4-cyclohexene-1,2-dicarboxylate

  • MDL number MFCD00059565
  • Cas Number : 2915-49-3
  • Molecular Weight : 394.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5303

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5303
Availability details for
Quantity
  • Availability
  • Add to Cart