COM638466603 Combi-Blocks, Inc.

Sodium 2-(dimethylamino)-1-hydroxyethanesulfonate

  • MDL number MFCD26743557
  • Cas Number : 1433960-19-0
  • Molecular Weight : 191.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5563

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5563
Availability details for
Quantity
  • Availability
  • Add to Cart