COM638466687 Combi-Blocks, Inc.

(5-Chloro-1-methyl-1h-benzo[d]imidazol-2-yl)methanol

  • MDL number MFCD02717142
  • Cas Number : 380177-22-0
  • Molecular Weight : 196.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5897

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5897
Availability details for
Quantity
  • Availability
  • Add to Cart