COM638466705 Combi-Blocks, Inc.

2-(2-Acetyl-4-bromophenoxy)acetic acid

  • MDL number MFCD27949767
  • Cas Number : 34849-51-9
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5962

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5962
Availability details for
Quantity
  • Availability
  • Add to Cart