COM638466858 Combi-Blocks, Inc.

3,9-Bis(3-aminopropyl)-2,4,8,10-tetraoxaspiro[5.5]undecane

  • MDL number MFCD00023818
  • Cas Number : 21587-74-6
  • Molecular Weight : 274.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6532
Availability details for
Quantity
  • Availability
  • Add to Cart