COM638466870 Combi-Blocks, Inc.

5-(tert-Butyl)isoindolin-1-one

  • MDL number MFCD24387350
  • Cas Number : 1361386-54-0
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6577
Availability details for
Quantity
  • Availability
  • Add to Cart