COM638466873 Combi-Blocks, Inc.

Ethyl 3-(6-aminopyridin-3-yl)propanoate

  • MDL number MFCD16036459
  • Cas Number : 666721-07-9
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6585

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6585
Availability details for
Quantity
  • Availability
  • Add to Cart