COM638466876 Combi-Blocks, Inc.

(S)-2-Amino-4-methyl-1-((r)-2-methyloxiran-2-yl)pentan-1-one 2,2,2-trifluoroacetate

  • MDL number MFCD28100284
  • Cas Number : 247068-85-5
  • Molecular Weight : 285.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6599

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6599
Availability details for
Quantity
  • Availability
  • Add to Cart