COM638466895 Combi-Blocks, Inc.

5-Bromo-2-methylthiazole-4-carbaldehyde

  • MDL number MFCD05156510
  • Cas Number : 221558-28-7
  • Molecular Weight : 206.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6683

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6683
Availability details for
Quantity
  • Availability
  • Add to Cart