COM638466936 Combi-Blocks, Inc.

Tributyl-n-octylphosphonium bromide

  • MDL number MFCD04038147
  • Cas Number : 57702-65-5
  • Molecular Weight : 395.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6826
Availability details for
Quantity
  • Availability
  • Add to Cart