COM638467043 Combi-Blocks, Inc.

2-Ethylhexyl 4-hydroxybenzoate

  • MDL number MFCD00016472
  • Cas Number : 5153-25-3
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7245

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7245
Availability details for
Quantity
  • Availability
  • Add to Cart