COM638467122 Combi-Blocks, Inc.

2,3,4,5-Tetrahydro-1h-benzo[e][1,4]diazepin-7-ylamine 2hcl

  • MDL number MFCD29059017
  • Cas Number : 1956380-40-7
  • Molecular Weight : 236.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7575
Availability details for
Quantity
  • Availability
  • Add to Cart