COM638467206 Combi-Blocks, Inc.

2-Chloro-3-iodobenzoic acid

  • MDL number MFCD06808822
  • Cas Number : 874817-93-3
  • Molecular Weight : 282.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7888
Availability details for
Quantity
  • Availability
  • Add to Cart