COM638467305 Combi-Blocks, Inc.

2-(Aminosulfonyl)-4-iodobenzoic acid methyl ester

  • MDL number MFCD11846230
  • Cas Number : 144550-79-8
  • Molecular Weight : 341.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8305

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8305
Availability details for
Quantity
  • Availability
  • Add to Cart