COM638467309 Combi-Blocks, Inc.

Pyridazine-3-carboxamide

  • MDL number MFCD19204130
  • Cas Number : 5450-54-4
  • Molecular Weight : 123.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8325

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8325
Availability details for
Quantity
  • Availability
  • Add to Cart