COM638467414 Combi-Blocks, Inc.

Terephthalic acid diisopropyl ester

  • MDL number MFCD01682720
  • Cas Number : 6422-84-0
  • Molecular Weight : 250.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8679

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8679
Availability details for
Quantity
  • Availability
  • Add to Cart