COM638467494 Combi-Blocks, Inc.

(3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl)methanol

  • MDL number MFCD22208006
  • Cas Number : 871727-08-1
  • Molecular Weight : 203.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8979

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-8979
Availability details for
Quantity
  • Availability
  • Add to Cart