COM638467505 Combi-Blocks, Inc.

N-Methyldodecanamide

  • MDL number MFCD00051780
  • Cas Number : 27563-67-3
  • Molecular Weight : 213.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9026

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9026
Availability details for
Quantity
  • Availability
  • Add to Cart