COM638467559 Combi-Blocks, Inc.

Labetalol

  • MDL number MFCD00242941
  • Cas Number : 36894-69-6
  • Molecular Weight : 328.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9252

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9252
Availability details for
Quantity
  • Availability
  • Add to Cart