COM638467597 Combi-Blocks, Inc.

1,3-Bis[1-(2-hydroxyethyl)-4-piperidyl]propane

  • MDL number MFCD00023767
  • Cas Number : 18073-84-2
  • Molecular Weight : 298.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9414

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9414
Availability details for
Quantity
  • Availability
  • Add to Cart