COM638467925 Combi-Blocks, Inc.

tert-Butyl 3-(6-bromopyridin-3-yl)propanoate

  • MDL number MFCD29059275
  • Cas Number : 1823119-39-6
  • Molecular Weight : 286.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QP-0776

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QP-0776
Availability details for
Quantity
  • Availability
  • Add to Cart